Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109486.1 | NP_001102956.1 | 7 | 554 |
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109484.1 | NP_001102954.1 | 6 | 609 | ||
XM_008765039.1 | XP_008763261.1 | 5 | 1635 | |||
XM_008765040.1 | XP_008763262.1 | 7 | 648 | |||
GenBank mRNA | BC166586.1 | - | 5 | 1606 | 7934414 | |
FQ217612.1 | - | 6 | 570 | |||
FQ217952.1 | - | 6 | 582 | |||
FQ221005.1 | - | 6 | 580 | |||
FQ221192.1 | - | 6 | 576 | |||
FQ221499.1 | - | 6 | 578 | |||
FQ221709.1 | - | 6 | 610 | |||
FQ221718.1 | - | 6 | 581 | |||
FQ221751.1 | - | 6 | 577 | |||
FQ221963.1 | - | 6 | 578 | |||
FQ222034.1 | - | 6 | 569 | |||
FQ222160.1 | - | 6 | 579 | |||
FQ222384.1 | - | 6 | 581 | |||
FQ222504.1 | - | 6 | 558 | |||
FQ223317.1 | - | 6 | 584 | |||
FQ228533.1 | - | 6 | 581 |
Gene Symbol: Myl6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100983.1 | NP_001094453.1 | 5 | 553 | ||
GenBank mRNA | S77858.1 | - | 5 | 553 |
Gene Symbol: RGD1559821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249290.2 | XP_006249352.1 | 1 | 575 |