Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134844.1 | NP_001128316.1 | 1784 | |||
XM_008765491.1 | XP_008763713.1 | 1827 | ||||
XM_008765492.1 | XP_008763714.1 | 1803 | ||||
XM_008765493.1 | XP_008763715.1 | 2189 | ||||
GenBank mRNA | BC166502.1 | - | 1784 | 7122207 | ||
K02428.1 | - | 150 |
Gene Symbol: Cyp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198676.1 | NP_001185605.1 | 9 | 1791 | ||
GenBank mRNA | K01626.1 | - | 2 | 615 | ||
K01721.1 | - | 6 | 1267 | |||
M13650.1 | - | 1 | 393 | |||
M34452.1 | - | 8 | 1480 |