Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49si2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009498.1 | NP_001009498.1 | 642 | |||
XM_008763406.1 | XP_008761628.1 | 655 | ||||
XM_008763407.1 | XP_008761629.1 | 655 | ||||
XM_008763408.1 | XP_008761630.1 | 655 | ||||
XM_008763409.1 | XP_008761631.1 | 655 | ||||
GenBank mRNA | AY659933.1 | - | 642 |
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172088.1 | NP_001165559.1 | 5 | 645 |
Gene Symbol: LOC103690111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237119.2 | XP_006237181.1 | 6 | 821 | ||
XM_006237121.2 | XP_006237183.1 | 6 | 821 |
Gene Symbol: LOC690097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763433.1 | XP_008761655.1 | 6 | 1068 |
Gene Symbol: Ly49si3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009919.1 | NP_001009919.1 | 5 | 642 | ||
XM_006237116.2 | XP_006237178.1 | 736 | ||||
XM_008763410.1 | XP_008761632.1 | 6 | 923 | |||
XM_008763411.1 | XP_008761633.1 | 6 | 1560 | |||
XM_008763412.1 | XP_008761634.1 | 6 | 717 | |||
XM_008763413.1 | XP_008761635.1 | 6 | 923 | |||
XM_008763414.1 | XP_008761636.1 | 6 | 923 | |||
XM_008763415.1 | XP_008761637.1 | 6 | 923 | |||
GenBank mRNA | AY653730.1 | - | 5 | 642 |