Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49si2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009498.1 | NP_001009498.1 | 303 | |||
XM_008763406.1 | XP_008761628.1 | 316 | ||||
XM_008763407.1 | XP_008761629.1 | 316 | ||||
XM_008763408.1 | XP_008761630.1 | 316 | ||||
XM_008763409.1 | XP_008761631.1 | 316 | ||||
GenBank mRNA | AY659933.1 | - | 303 |
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172088.1 | NP_001165559.1 | 3 | 306 |
Gene Symbol: LOC100910264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763346.1 | XP_008761568.1 | 3 | 2882 |
Gene Symbol: LOC100910679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763462.1 | XP_008761684.1 | 3 | 318 |
Gene Symbol: LOC502907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763434.1 | XP_008761656.1 | 4 | 985 |
Gene Symbol: Ly49si1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009497.1 | NP_001009497.1 | 3 | 306 | ||
XM_008763312.1 | XP_008761534.1 | 3 | 557 | |||
XM_008763313.1 | XP_008761535.1 | 4 | 587 | |||
XM_008763314.1 | XP_008761536.1 | 4 | 381 | |||
XM_008763315.1 | XP_008761537.1 | 3 | 557 | |||
XM_008763316.1 | XP_008761538.1 | 3 | 557 | |||
XM_008763317.1 | XP_008761539.1 | 2 | 467 | |||
XM_008763318.1 | XP_008761540.1 | 2 | 467 | |||
XM_008763319.1 | XP_008761541.1 | 3 | 557 | |||
GenBank mRNA | AY659935.1 | - | 3 | 306 |
Gene Symbol: Ly49si3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009919.1 | NP_001009919.1 | 3 | 303 | ||
XM_006237116.2 | XP_006237178.1 | 397 | ||||
XM_008763410.1 | XP_008761632.1 | 4 | 584 | |||
XM_008763411.1 | XP_008761633.1 | 4 | 1221 | |||
XM_008763412.1 | XP_008761634.1 | 4 | 378 | |||
XM_008763413.1 | XP_008761635.1 | 4 | 584 | |||
XM_008763414.1 | XP_008761636.1 | 4 | 584 | |||
XM_008763415.1 | XP_008761637.1 | 4 | 584 | |||
GenBank mRNA | AY653730.1 | - | 3 | 303 |