Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 16 | 2382 | ||
NM_001300952.1 | NP_001287881.1 | 16 | 2382 | |||
NM_015326.4 | NP_056141.2 | 16 | 2385 | |||
XM_005277510.2 | XP_005277567.1 | 17 | 2572 | |||
XM_005277511.2 | XP_005277568.1 | 17 | 2569 | |||
XM_005277514.1 | XP_005277571.1 | 17 | 2572 | |||
XM_005277515.1 | XP_005277572.1 | 17 | 2569 | |||
XM_011509354.1 | XP_011507656.1 | 17 | 2572 | |||
XM_011509355.1 | XP_011507657.1 | 17 | 2569 | |||
XM_011509356.2 | XP_011507658.1 | 17 | 2572 | |||
XM_011509359.1 | XP_011507661.1 | 17 | 2572 | |||
XM_017000838.1 | XP_016856327.1 | 15 | 2216 | |||
XM_017000839.1 | XP_016856328.1 | 15 | 2246 | |||
XM_017000840.1 | XP_016856329.1 | 17 | 1861 | |||
XM_017000841.1 | XP_016856330.1 | 17 | 2569 | |||
GenBank mRNA | AB007925.1 | - | 16 | 1915 | ||
AK293335.1 | - | 13 | 1421 | |||
AK295845.1 | - | 16 | 2138 | |||
BC041635.1 | - | 8 | 686 | |||
BC132872.1 | - | 16 | 1913 | 40146894 | ||
BC132874.1 | - | 16 | 1913 | 40146896 | ||
BC144343.1 | - | 16 | 1910 | 9052863 |