Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135007.1 | NP_001128479.1 | 3, 4 | 819 | ||
XM_006241885.2 | XP_006241947.1 | 3, 4 | 814 |