Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC689770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142304.1 | NP_001135776.1 | 410 | |||
GenBank mRNA | EU128747.1 | - | 416 | |||
FJ404474.1 | - | 410 |
Gene Symbol: LOC689757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101019.2 | NP_001094489.2 | 3 | 440 | ||
XM_008763332.1 | XP_008761554.1 | 3 | 743 | |||
GenBank mRNA | EF100687.1 | - | 3 | 410 | ||
FJ404472.1 | - | 3 | 440 |