Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC689770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142304.1 | NP_001135776.1 | 737 | |||
GenBank mRNA | EU128747.1 | - | 743 | |||
FJ404474.1 | - | 737 |