Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134705.1 | NP_001128177.1 | 2 | 342 | ||
GenBank mRNA | FQ217721.1 | - | 2 | 291 | ||
FQ217795.1 | - | 2 | 291 | |||
FQ221226.1 | - | 2 | 352 | |||
FQ223619.1 | - | 2 | 293 | |||
FQ224715.1 | - | 2 | 321 |