Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPECC1L-ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103546.1 | 3 | 362 | |||
GenBank mRNA | AB209022.1 | - | 3 | 281 |
Gene Symbol: SPECC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145468.3 | NP_001138940.3 | 2 | 258 | ||
NM_001254732.2 | NP_001241661.2 | 2 | 258 | |||
NM_015330.4 | NP_056145.4 | 3 | 362 | |||
GenBank mRNA | AK097129.1 | - | 2 | 246 | ||
AK301482.1 | - | 2 | 193 | |||
AL832425.1 | - | 3 | 281 | |||
AY884293.1 | - | 2 | 186 | |||
BC005093.1 | - | 3 | 281 | |||
BC136471.1 | - | 2 | 128 | 9020460 | ||
DA385554.1 | - | 2 | 252 |