Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290184.1 | NP_001277113.1 | 12 | 1556 | ||
NM_001290186.1 | NP_001277115.1 | 8 | 1003 | |||
NM_015331.2 | NP_056146.1 | 11 | 1417 | |||
XM_011509363.2 | XP_011507665.1 | 11 | 1634 | |||
GenBank mRNA | AF240468.1 | - | 11 | 1435 | ||
AK296153.1 | - | 6 | 702 | |||
AK299142.1 | - | 8 | 890 | |||
AK314764.1 | - | 11 | 1324 | |||
AY359120.1 | - | 11 | 1294 | |||
BC047621.1 | - | 12 | 1513 | 4827966 | ||
D87442.1 | - | 11 | 1291 |