Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015332.3 | NP_056147.2 | 2 | 556 | ||
XM_011515247.2 | XP_011513549.1 | 2 | 645 | |||
XM_017011908.1 | XP_016867397.1 | 2 | 645 | |||
XM_017011909.1 | XP_016867398.1 | 2 | 412 | |||
XM_017011910.1 | XP_016867399.1 | 2 | 270 | |||
GenBank mRNA | AB028991.1 | - | 2 | 350 | ||
AK024226.1 | - | 1 | 64 | |||
AK307613.1 | - | 1 | 77 | |||
BC003691.2 | - | 2 | 417 | 3453305 | ||
BC011673.2 | - | 1 | 176 | |||
BC035014.2 | - | 2 | 454 | 4812229 | ||
CN386226.1 | - | 2 | 556 | |||
EU350932.1 | - | 2 | 374 |