Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001745.3 | NP_001736.1 | 4 | 924 | ||
XM_011543653.1 | XP_011541955.1 | 3 | 1411 | |||
GenBank mRNA | AF068179.1 | - | 4 | 641 | ||
AK313156.1 | - | 4 | 848 | |||
BC063532.1 | - | 3 | 360 | |||
BC130325.1 | - | 4 | 856 | 40146356 | ||
U18242.1 | - | 4 | 819 |