Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025164.4 | NP_079440.3 | 19, 20 | 2629 | ||
XM_005271481.2 | XP_005271538.1 | 20, 21 | 2811 | |||
XM_005271482.4 | XP_005271539.2 | 19, 20 | 2667 | |||
XM_005271485.3 | XP_005271542.1 | 7, 8 | 1149 | |||
XM_011542721.2 | XP_011541023.1 | 20, 21 | 2773 | |||
XM_011542723.2 | XP_011541025.1 | 20, 21 | 2654 | |||
XM_011542724.2 | XP_011541026.1 | 19, 20 | 2508 | |||
XM_011542725.2 | XP_011541027.1 | 18, 19 | 2391 | |||
XM_017017425.1 | XP_016872914.1 | 20, 21 | 2849 | |||
XM_017017426.1 | XP_016872915.1 | 18, 19 | 2723 | |||
XM_017017427.1 | XP_016872916.1 | 15, 16 | 2126 | |||
GenBank mRNA | AB209327.1 | - | 12, 13 | 1639 | ||
AK022302.1 | - | 1, 2 | 184 | |||
AY598338.1 | - | 1, 2 | 181 | |||
BC035583.2 | - | 2, 3 | 459 | |||
BC086858.1 | - | 1, 2 | 184 |