Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015335.4 | NP_056150.1 | 18 | 4022 | ||
XM_011538080.2 | XP_011536382.1 | 18 | 4055 | |||
XM_011538081.2 | XP_011536383.1 | 18 | 4052 | |||
XM_011538082.2 | XP_011536384.1 | 18 | 4225 | |||
XM_017019090.1 | XP_016874579.1 | 18 | 4052 | |||
GenBank mRNA | AB028948.2 | - | 12 | 3089 | ||
AF515599.1 | - | 18 | 4022 | |||
AK023837.1 | - | 2 | 739 | |||
AK127706.1 | - | 15 | 3556 | |||
AL133033.1 | - | 12 | 3062 | |||
AY338463.1 | - | 18 | 4174 | |||
BC130422.1 | - | 18 | 4018 | 40146453 | ||
CR749332.1 | - | 16 | 3546 |