Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015335.4 | NP_056150.1 | 21 | 4628 | ||
XM_011538080.2 | XP_011536382.1 | 21 | 4661 | |||
XM_011538081.2 | XP_011536383.1 | 21 | 4658 | |||
XM_011538082.2 | XP_011536384.1 | 21 | 4831 | |||
XM_017019090.1 | XP_016874579.1 | 21 | 4658 | |||
GenBank mRNA | AB028948.2 | - | 15 | 3695 | ||
AF515599.1 | - | 21 | 4628 | |||
AK023837.1 | - | 5 | 1345 | |||
AL133033.1 | - | 15 | 3668 | |||
AY338463.1 | - | 21 | 4780 | |||
BC130422.1 | - | 21 | 4624 | 40146453 | ||
CR749332.1 | - | 19 | 4152 |