Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 5 | 680 | ||
XM_005268750.4 | XP_005268807.1 | 5 | 520 | |||
XM_011538083.1 | XP_011536385.1 | 5 | 544 | |||
XM_017019091.1 | XP_016874580.1 | 5 | 520 | |||
XM_017019092.1 | XP_016874581.1 | 4 | 424 | |||
XM_017019093.1 | XP_016874582.1 | 4 | 373 | |||
XM_017019094.1 | XP_016874583.1 | 4 | 401 | |||
GenBank mRNA | AB023163.1 | - | 5 | 624 | ||
AB097013.1 | - | 5 | 500 | |||
AB097035.1 | - | 5 | 500 | |||
AF049612.1 | - | 5 | 513 | |||
AF161412.1 | - | 5 | 512 | |||
AK292866.1 | - | 5 | 496 | |||
AK299089.1 | - | 4 | 392 | |||
BC030990.1 | - | 4 | 547 | |||
BC050324.1 | - | 5 | 639 | 5266097 |