Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 1 | 216 | ||
XM_005268750.4 | XP_005268807.1 | 1 | 56 | |||
XM_017019091.1 | XP_016874580.1 | 1 | 56 | |||
XM_017019093.1 | XP_016874582.1 | 1 | 13 | |||
XM_017019094.1 | XP_016874583.1 | 1 | 41 | |||
GenBank mRNA | AB023163.1 | - | 1 | 160 | ||
AB024494.1 | - | 1 | 703 | |||
AB097013.1 | - | 1 | 36 | |||
AB097035.1 | - | 1 | 36 | |||
AF049612.1 | - | 1 | 49 | |||
AF161412.1 | - | 1 | 48 | |||
AK292866.1 | - | 1 | 32 | |||
AK299089.1 | - | 1 | 32 | |||
BC030990.1 | - | 1 | 187 | |||
BC050324.1 | - | 1 | 175 | 5266097 | ||
DA227374.1 | - | 1 | 216 |