Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080398.1 | NP_001073867.1 | 44 | 5208 | ||
XM_005251850.3 | XP_005251907.1 | 42 | 4711 | |||
XM_005251851.4 | XP_005251908.1 | 43 | 5090 | |||
XM_005251852.4 | XP_005251909.1 | 43 | 5042 | |||
XM_005251853.3 | XP_005251910.1 | 43 | 4824 | |||
XM_005251854.3 | XP_005251911.1 | 42 | 4993 | |||
XM_011518422.2 | XP_011516724.1 | 42 | 4711 | |||
XM_017014557.1 | XP_016870046.1 | 36 | 4030 | |||
GenBank mRNA | AB002366.1 | - | 32 | 3482 | ||
AK024705.1 | - | 19 | 2037 | |||
AK025689.1 | - | 27 | 3010 | |||
AK027063.1 | - | 12 | 1178 | |||
AK127247.1 | - | 38 | 4149 | |||
BC021127.1 | - | 8 | 792 |