Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 16 | 2222 | ||
XM_005263565.4 | XP_005263622.1 | 16 | 2357 | |||
XM_005263568.4 | XP_005263625.1 | 16 | 2357 | |||
XM_011510490.2 | XP_011508792.1 | 16 | 2257 | |||
XM_017003164.1 | XP_016858653.1 | 16 | 2227 | |||
XM_017003165.1 | XP_016858654.1 | 17 | 2326 | |||
GenBank mRNA | AK294196.1 | - | 8 | 668 | ||
AK308695.1 | - | 8 | 670 | |||
AK316584.1 | - | 16 | 2229 | |||
BC114917.1 | - | 16 | 1936 | 40068588 | ||
BC114918.1 | - | 16 | 1936 | 40012335 | ||
D16316.1 | - | 3 | 246 | |||
U08023.1 | - | 16 | 2237 |