Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015340.3 | NP_056155.1 | 16, 17 | 2044 | ||
XM_005265006.1 | XP_005265063.1 | 15, 16 | 2000 | |||
XM_011533554.2 | XP_011531856.1 | 16, 17 | 2063 | |||
XM_017006042.1 | XP_016861531.1 | 16, 17 | 2063 | |||
GenBank mRNA | AK025465.1 | - | 10, 11 | 1344 | ||
AK026113.1 | - | 3, 4 | 267 | |||
AK308146.1 | - | 14, 15 | 1868 | |||
BC025989.2 | - | 16, 17 | 2051 | 4822546 | ||
D21851.1 | - | 16, 17 | 2044 |