Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278926.1 | NP_001265855.1 | 3 | 329 | ||
NM_001278927.1 | NP_001265856.1 | 2 | 292 | |||
NM_015342.3 | NP_056157.1 | 2 | 295 | |||
XM_006714583.3 | XP_006714646.1 | 2 | 188 | |||
XM_017009286.1 | XP_016864775.1 | 2 | 67 | |||
GenBank mRNA | AK025679.1 | - | 2 | 242 | ||
AK298175.1 | - | 2 | 232 | |||
AK300022.1 | - | 3 | 272 | |||
BC015385.1 | - | 2 | 223 | 4390706 | ||
BX642958.1 | - | 2 | 300 | |||
D38552.1 | - | 2 | 218 | |||
DB454273.1 | - | 2 | 292 |