Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDNEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143775.1 | NP_001137247.1 | 3, 4 | 669 | ||
NM_015343.4 | NP_056158.2 | 4, 5 | 736 | |||
GenBank mRNA | AJ011916.1 | - | 3, 4 | 319 | ||
AY364239.1 | - | 3, 4 | 289 | |||
BC009295.2 | - | 4, 5 | 735 | 4123279 |