Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 7 | 689 | ||
NM_001746.3 | NP_001737.1 | 7 | 742 | |||
XM_011534664.1 | XP_011532966.1 | 7 | 1107 | |||
XM_011534665.2 | XP_011532967.1 | 7 | 715 | |||
GenBank mRNA | AJ271880.1 | - | 6 | 581 | ||
AK129990.1 | - | 5 | 449 | |||
AK294702.1 | - | 7 | 745 | |||
AK304420.1 | - | 6 | 624 | |||
AK312334.1 | - | 7 | 649 | |||
BC003552.1 | - | 7 | 676 | 3546389 | ||
BC042843.1 | - | 7 | 623 | 4342723 | ||
L10284.1 | - | 7 | 654 | |||
L18887.1 | - | 7 | 660 | |||
M94859.1 | - | 7 | 660 | |||
S67659.1 | - | 6 | 517 |