Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 3 | 497 | ||
NM_012239.5 | NP_036371.1 | 3 | 643 | |||
XM_005252835.1 | XP_005252892.1 | 3 | 643 | |||
XM_011519956.1 | XP_011518258.1 | 3 | 1435 | |||
XM_011519957.1 | XP_011518259.1 | 3 | 1264 | |||
XM_017017428.1 | XP_016872917.1 | 3 | 489 | |||
XM_017017429.1 | XP_016872918.1 | 3 | 663 | |||
XM_017017430.1 | XP_016872919.1 | 3 | 657 | |||
XM_017017431.1 | XP_016872920.1 | 3 | 388 | |||
GenBank mRNA | AF083108.2 | - | 3 | 711 | ||
AK299009.1 | - | 3 | 633 | |||
AK299438.1 | - | 3 | 475 | |||
AK301558.1 | - | 2 | 443 | |||
AK301979.1 | - | 3 | 631 | |||
AK308681.1 | - | 3 | 644 | |||
AK316175.1 | - | 3 | 547 | |||
AL535769.3 | - | 3 | 478 | |||
BC001042.2 | - | 3 | 626 | 2823471 | ||
BI755839.1 | - | 3 | 443 |