Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 11 | 1449 | ||
NM_001746.3 | NP_001737.1 | 11 | 1502 | |||
XM_011534664.1 | XP_011532966.1 | 11 | 1867 | |||
XM_011534665.2 | XP_011532967.1 | 11 | 1475 | |||
GenBank mRNA | AJ271880.1 | - | 10 | 1341 | ||
AK294702.1 | - | 11 | 1505 | |||
AK304420.1 | - | 10 | 1384 | |||
AK312334.1 | - | 11 | 1409 | |||
BC003552.1 | - | 11 | 1436 | 3546389 | ||
BC042843.1 | - | 11 | 1383 | 4342723 | ||
L10284.1 | - | 11 | 1414 | |||
L18887.1 | - | 11 | 1420 | |||
M94859.1 | - | 11 | 1420 | |||
M98452.1 | - | 4 | 617 |