Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD3-BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204062.1 | NP_001190991.1 | 3 | 334 | ||
GenBank mRNA | AK313235.1 | - | 3 | 309 |
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012071.3 | NP_036203.1 | 3 | 334 | ||
GenBank mRNA | AF078848.1 | - | 3 | 309 | ||
AF201948.1 | - | 3 | 328 | |||
AK128789.1 | - | 3 | 308 | |||
AY542159.1 | - | 3 | 299 | |||
BC022898.2 | - | 3 | 321 | 3531636 | ||
BF695081.1 | - | 335 | ||||
BG496810.1 | - | 3 | 328 | |||
BX647275.1 | - | 3 | 331 |