Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 2 | 189 | ||
NM_001746.3 | NP_001737.1 | 2 | 242 | |||
XM_011534664.1 | XP_011532966.1 | 2 | 607 | |||
XM_011534665.2 | XP_011532967.1 | 2 | 215 | |||
GenBank mRNA | AJ271880.1 | - | 1 | 81 | ||
AK129990.1 | - | 2 | 141 | |||
AK294702.1 | - | 2 | 245 | |||
AK304420.1 | - | 2 | 183 | |||
AK312334.1 | - | 2 | 149 | |||
BC003552.1 | - | 2 | 176 | 3546389 | ||
BC042843.1 | - | 2 | 123 | 4342723 | ||
L10284.1 | - | 2 | 154 | |||
L18887.1 | - | 2 | 160 | |||
M94859.1 | - | 2 | 160 | |||
S67659.1 | - | 1 | 20 |