Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLYCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012213.2 | NP_036345.2 | 5 | 1081 | ||
GenBank mRNA | AF090834.1 | - | 5 | 1061 | ||
AF097832.2 | - | 5 | 999 | |||
AF153679.1 | - | 5 | 1039 | |||
AK094095.1 | - | 1 | 2004 | |||
BC000286.1 | - | 5 | 1033 | 3357140 | ||
BC052592.1 | - | 5 | 1081 | 6421051 |