Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 19 | 2253 | ||
GenBank mRNA | BC065535.1 | - | 19 | 2206 | 6007603 | |
X57398.1 | - | 19 | 2091 |