Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178074.1 | NP_001171545.1 | 16 | 2164 | ||
NM_021150.3 | NP_066973.2 | 16 | 2164 | |||
XM_005268754.4 | XP_005268811.1 | 17 | 2240 | |||
XM_005268757.4 | XP_005268814.1 | 16 | 2159 | |||
XM_011538093.2 | XP_011536395.1 | 17 | 2293 | |||
XM_017019098.1 | XP_016874587.1 | 17 | 2589 | |||
XM_017019099.1 | XP_016874588.1 | 17 | 5248 | |||
XM_017019100.1 | XP_016874589.1 | 16 | 2433 | |||
XM_017019101.1 | XP_016874590.1 | 16 | 5092 | |||
XM_017019102.1 | XP_016874591.1 | 16 | 5089 | |||
GenBank mRNA | AJ133439.1 | - | 10 | 1287 | ||
AK095519.1 | - | 16 | 2164 | |||
AK295565.1 | - | 15 | 1970 | |||
AK307576.1 | - | 16 | 2002 | |||
BC029871.1 | - | 14 | 1728 | |||
BC115393.1 | - | 16 | 1992 | 40053971 | ||
BC115394.1 | - | 16 | 1992 | 40053972 |