Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267036.1 | NP_001253965.1 | 4 | 608 | ||
NM_001267037.1 | NP_001253966.1 | 3 | 353 | |||
NM_012244.3 | NP_036376.2 | 6 | 1569 | |||
NM_182728.2 | NP_877392.1 | 4 | 507 | |||
GenBank mRNA | AB037669.1 | - | 6 | 1532 | ||
AF087908.1 | - | 4 | 462 | |||
AF087911.1 | - | 5 | 618 | |||
AF135828.1 | - | 6 | 1044 | |||
AF135830.1 | - | 2 | 150 | |||
AF171669.1 | - | 6 | 1046 | |||
AK223610.1 | - | 6 | 1568 | |||
AK296702.1 | - | 4 | 561 | |||
AK300384.1 | - | 3 | 296 | |||
AK307609.1 | - | 4 | 441 | |||
AK313465.1 | - | 6 | 823 | |||
AL365343.2 | - | 6 | 1073 | |||
AL365345.1 | - | 2 | 186 | |||
AL365414.1 | - | 6 | 1073 | |||
BC052250.1 | - | 6 | 852 | 6260264 | ||
BX248288.1 | - | 4 | 458 | |||
DC348596.1 | - | 3 | 353 | |||
Y18483.1 | - | 6 | 1553 |