Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267036.1 | NP_001253965.1 | 3 | 560 | ||
NM_012244.3 | NP_036376.2 | 5 | 1521 | |||
NM_182728.2 | NP_877392.1 | 3 | 459 | |||
NR_049767.1 | 3 | 560 | ||||
GenBank mRNA | AB037669.1 | - | 5 | 1484 | ||
AF087908.1 | - | 3 | 414 | |||
AF087911.1 | - | 4 | 570 | |||
AF135828.1 | - | 5 | 996 | |||
AF135830.1 | - | 1 | 102 | |||
AF171669.1 | - | 5 | 998 | |||
AK223610.1 | - | 5 | 1520 | |||
AK296702.1 | - | 3 | 513 | |||
AK307609.1 | - | 3 | 393 | |||
AK313465.1 | - | 5 | 775 | |||
AL365342.1 | - | 5 | 1506 | |||
AL365343.2 | - | 5 | 1025 | |||
AL365345.1 | - | 1 | 138 | |||
AL365414.1 | - | 5 | 1025 | |||
BC036825.1 | - | 3 | 411 | |||
BC052250.1 | - | 5 | 804 | 6260264 | ||
BX248288.1 | - | 3 | 410 | |||
Y18483.1 | - | 5 | 1505 |