Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252127.1 | NP_001239056.1 | 18 | 2716 | ||
NM_007347.4 | NP_031373.2 | 18 | 2693 | |||
XM_005254264.3 | XP_005254321.1 | 18 | 2511 | |||
XM_006720447.3 | XP_006720510.1 | 18 | 2871 | |||
XM_017022042.1 | XP_016877531.1 | 18 | 2726 | |||
GenBank mRNA | AB030653.1 | - | 18 | 2616 | ||
AF155156.2 | - | 18 | 2619 | |||
BC126308.1 | - | 18 | 2692 | 8992024 | ||
BC130466.1 | - | 18 | 2692 | 40146497 | ||
CR749604.1 | - | 18 | 2593 |