Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 5 | 614 | ||
GenBank mRNA | AK092382.1 | - | 4, 5 | 286 | ||
AK315798.1 | - | 5 | 483 | |||
BC005216.1 | - | 5 | 488 | 3949903 | ||
BI760781.1 | - | 5 | 613 | |||
BM129235.1 | - | 4 | 394 | |||
M16630.1 | - | 5 | 459 | |||
M18692.1 | - | 5 | 486 |