Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012098.2 | NP_036230.1 | 2 | 615 | ||
XM_006717030.3 | XP_006717093.1 | 2 | 653 | |||
GenBank mRNA | AF125175.1 | - | 1 | 136 | ||
AK307175.1 | - | 2 | 511 | |||
AK312876.1 | - | 2 | 534 | |||
AL598400.1 | - | 2 | 616 | |||
AY358274.1 | - | 2 | 634 | |||
BC012368.1 | - | 2 | 488 | 3865062 |