Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243013.1 | NP_001229942.1 | 11 | 2207 | ||
NM_019624.3 | NP_062570.1 | 11 | 2267 | |||
NM_019625.3 | NP_062571.1 | 12 | 2396 | |||
XM_005253558.2 | XP_005253615.1 | 12 | 2336 | |||
XM_011538095.2 | XP_011536397.1 | 12 | 2619 | |||
XM_011538096.2 | XP_011536398.1 | 12 | 2415 | |||
XM_011538098.2 | XP_011536400.1 | 11 | 3261 | |||
XM_017019103.1 | XP_016874592.1 | 13 | 2457 | |||
XM_017019104.1 | XP_016874593.1 | 10 | 1755 | |||
GenBank mRNA | AB040953.2 | - | 12 | 4925 | ||
AB045381.2 | - | 11 | 2145 | |||
AB177852.1 | - | 12 | 2308 | |||
AF216494.1 | - | 12 | 2384 | |||
AK074738.1 | - | 9 | 1340 | |||
AK294225.1 | - | 6 | 955 | |||
AK304295.1 | - | 11 | 2203 | |||
BC064384.1 | - | 12 | 2673 | 5744451 | ||
U66676.1 | - | 3 | 232 |