Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003962.2 | NP_001003962.1 | 4 | 549 | ||
NM_001302632.1 | NP_001289561.1 | 4 | 456 | |||
NM_001302633.1 | NP_001289562.1 | 3 | 241 | |||
NM_001749.3 | NP_001740.1 | 4 | 552 | |||
XM_005259295.1 | XP_005259352.1 | 4 | 352 | |||
XM_005259296.1 | XP_005259353.1 | 4 | 355 | |||
XM_011527359.1 | XP_011525661.1 | 3 | 182 | |||
GenBank mRNA | AK289606.1 | - | 4 | 491 | ||
AY007141.1 | - | 4 | 349 | |||
BC000592.2 | - | 4 | 370 | 3347146 | ||
BC007779.2 | - | 4 | 380 | 4301394 | ||
BC011903.2 | - | 4 | 370 | 4120579 | ||
BC017308.2 | - | 4 | 349 | 5089022 | ||
BC018931.2 | - | 4 | 440 | 4303926 | ||
BC021933.1 | - | 4 | 344 | 4872635 | ||
BC023643.2 | - | 4 | 342 | 5017512 | ||
BC064998.1 | - | 4 | 457 | 5785274 | ||
BQ898664.1 | - | 4 | 462 | |||
BT009775.1 | - | 3 | 305 | |||
CN349928.1 | - | 3 | 190 | |||
HY003903.1 | - | 3 | 244 | |||
X04106.1 | - | 4 | 463 |