Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320867.1 | NP_001307796.1 | 5, 6 | 639 | ||
NM_001320868.1 | NP_001307797.1 | 4, 5 | 523 | |||
NM_001320869.1 | NP_001307798.1 | 3, 4 | 378 | |||
NM_014297.4 | NP_055112.2 | 5, 6 | 672 | |||
XM_005258687.3 | XP_005258744.1 | 5, 6 | 769 | |||
GenBank mRNA | AK309880.1 | - | 5, 6 | 617 | ||
AK315346.1 | - | 5, 6 | 635 | |||
BC008250.1 | - | 5, 6 | 622 | 3872059 | ||
BG706505.1 | - | 4, 5 | 472 | |||
D83198.2 | - | 5, 6 | 661 | |||
KU178565.1 | - | 5, 6 | 593 | |||
KU178566.1 | - | 5, 6 | 560 | |||
KU178567.1 | - | 3, 4 | 299 |