Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 2 | 1289 | ||
NM_001301141.1 | NP_001288070.1 | 2 | 1289 | |||
NM_001320042.1 | NP_001306971.1 | 2 | 1289 | |||
NM_014301.4 | NP_055116.1 | 3 | 1385 | |||
NM_213595.3 | NP_998760.1 | 2 | 1289 | |||
NR_135127.1 | 3 | 1385 | ||||
XM_011538100.2 | XP_011536402.1 | 3 | 1385 | |||
XM_011538101.2 | XP_011536403.1 | 2 | 1289 | |||
GenBank mRNA | AK000574.1 | - | 2 | 132 | ||
AK022758.1 | - | 1 | 1713 | |||
AK057251.1 | - | 2 | 1289 | |||
AK293202.1 | - | 2 | 142 | |||
AK297862.1 | - | 2 | 161 | |||
AY009127.1 | - | 3 | 242 | |||
AY009128.1 | - | 2 | 146 | |||
BC011906.2 | - | 2 | 145 | 4136262 | ||
BC061903.1 | - | 2 | 138 | 4183674 | ||
BG702203.1 | - | 2 | 140 | |||
BI597126.1 | - | 2 | 240 | |||
BM423850.1 | - | 2 | 135 | |||
BM921073.1 | - | 2 | 230 | |||
BP378051.1 | - | 2 | 161 | |||
BU602914.1 | - | 2 | 150 | |||
DB095495.1 | - | 2 | 156 | |||
DB493673.1 | - | 2 | 219 | |||
EU329002.1 | - | 3 | 242 |