Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012456.1 | NP_001012474.1 | 3 | 227 | ||
NM_014302.3 | NP_055117.1 | 3 | 231 | |||
GenBank mRNA | AF054184.1 | - | 3 | 226 | ||
AF086539.1 | - | 3 | 187 | |||
AK308617.1 | - | 3 | 179 | |||
AK311845.1 | - | 3 | 228 | |||
BC009480.2 | - | 3 | 169 | 3954682 | ||
BC051840.1 | - | 151 | 6060540 | |||
BU932672.1 | - | 230 | ||||
CR456979.1 | - | 2 | 136 |