Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128208.1 | NP_001121680.1 | 2 | 159 | ||
NM_015344.2 | NP_056159.2 | 2 | 159 | |||
GenBank mRNA | AF063605.1 | - | 2 | 136 | ||
AF161461.1 | - | 2 | 95 | |||
AK075363.1 | - | 2 | 103 | |||
AY277595.1 | - | 2 | 131 | |||
AY358674.1 | - | 2 | 131 | |||
BC000642.2 | - | 2 | 142 | 3344487 | ||
CR541735.1 | - | 1 | 52 | |||
CR541760.1 | - | 1 | 52 | |||
DA258969.1 | - | 2 | 159 |