Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201427.1 | NP_001188356.1 | 20 | 2618 | ||
NM_015345.3 | NP_056160.2 | 20 | 2682 | |||
XM_006715039.3 | XP_006715102.3 | 21 | 3290 | |||
XM_006715040.3 | XP_006715103.3 | 20 | 3262 | |||
XM_006715042.2 | XP_006715105.1 | 21 | 5285 | |||
XM_006715043.2 | XP_006715106.1 | 21 | 2662 | |||
XM_006715045.3 | XP_006715108.1 | 21 | 3877 | |||
XM_006715046.3 | XP_006715109.1 | 21 | 3055 | |||
XM_017010630.1 | XP_016866119.1 | 21 | 3901 | |||
GenBank mRNA | AB002379.2 | - | 20 | 2618 | ||
BX647839.1 | - | 4 | 3200 |