Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 34 | 6468 | ||
XM_017021124.1 | XP_016876613.1 | 34 | 6487 | |||
XM_017021125.1 | XP_016876614.1 | 34 | 6487 | |||
XM_017021126.1 | XP_016876615.1 | 33 | 7101 | |||
XM_017021127.1 | XP_016876616.1 | 23 | 4183 | |||
XM_017021128.1 | XP_016876617.1 | 23 | 4036 | |||
GenBank mRNA | AB002319.1 | - | 19 | 3338 | ||
AB425197.1 | - | 34 | 6456 | |||
AK024743.1 | - | 6 | 830 | |||
BC033235.1 | - | 1 | 163 | |||
BC144594.1 | - | 32 | 5772 | |||
BX537886.1 | - | 34 | 6462 | |||
BX538025.1 | - | 30 | 5961 | |||
BX648683.1 | - | 34 | 6466 | |||
CR749276.1 | - | 35 | 6403 | |||
CR749365.1 | - | 34 | 6467 |