Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 7 | 1817 | ||
NM_015349.2 | NP_056164.1 | 5 | 1550 | |||
GenBank mRNA | AK091434.1 | - | 5 | 1442 | ||
AK093526.1 | - | 5 | 2161 | |||
AK294706.1 | - | 5 | 1442 | |||
AL833540.1 | - | 5 | 1550 | |||
BC130302.1 | - | 5 | 1473 | 40146333 | ||
CR627378.1 | - | 6 | 1515 | |||
D87077.2 | - | 8 | 1714 |