Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015353.2 | NP_056168.1 | 3 | 584 | ||
NR_110834.1 | 4 | 465 | ||||
NR_110835.1 | 5 | 547 | ||||
XM_011524575.2 | XP_011522877.1 | 3 | 546 | |||
GenBank mRNA | AK307083.1 | - | 5 | 502 | ||
BC033329.1 | - | 4 | 465 | |||
BM906741.1 | - | 539 | ||||
D79998.1 | - | 3 | 523 |