Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUZ12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321207.1 | NP_001308136.1 | 5, 6 | 767 | ||
NM_015355.3 | NP_056170.2 | 6, 7 | 836 | |||
XM_006721794.3 | XP_006721857.1 | 4, 5 | 278 | |||
GenBank mRNA | AK074333.1 | - | 1, 2 | 73 | ||
AK290014.1 | - | 6, 7 | 807 | |||
BC015704.1 | - | 6, 7 | 678 | 3908024 | ||
BC018583.1 | - | 5, 6 | 710 | |||
D63881.2 | - | 6, 7 | 793 |