Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256468.1 | NP_001243397.1 | 13 | 2758 | ||
NM_001256469.1 | NP_001243398.1 | 13 | 2431 | |||
NM_012330.3 | NP_036462.2 | 13 | 3307 | |||
XM_005269664.2 | XP_005269721.1 | 13 | 3368 | |||
XM_017016000.1 | XP_016871489.1 | 13 | 3216 | |||
XM_017016001.1 | XP_016871490.1 | 13 | 3080 | |||
XM_017016002.1 | XP_016871491.1 | 13 | 3319 | |||
XM_017016003.1 | XP_016871492.1 | 12 | 2990 | |||
XM_017016004.1 | XP_016871493.1 | 12 | 3054 | |||
XM_017016005.1 | XP_016871494.1 | 13 | 2667 | |||
XM_017016006.1 | XP_016871495.1 | 13 | 2340 | |||
XM_017016007.1 | XP_016871496.1 | 13 | 2203 | |||
XM_017016008.1 | XP_016871497.1 | 12 | 2114 | |||
XM_017016009.1 | XP_016871498.1 | 12 | 2269 | |||
GenBank mRNA | AB002381.2 | - | 11 | 1730 | ||
AF113514.1 | - | 12 | 2045 | |||
AF119230.1 | - | 12 | 2372 | |||
AF119231.1 | - | 12 | 2921 | |||
AF217500.1 | - | 13 | 3094 | |||
AK074816.1 | - | 13 | 2244 | |||
AL832065.1 | - | 4 | 4559 | |||
BC150270.1 | - | 11 | 1757 | 9007238 | ||
BC150618.1 | - | 13 | 3067 | 9056988 |