Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 7 | 743 | ||
XM_005254228.3 | XP_005254285.1 | 5 | 965 | |||
XM_011521363.2 | XP_011519665.1 | 5 | 695 | |||
GenBank mRNA | AF137378.2 | - | 7 | 746 | ||
AK095863.1 | - | 8 | 755 |