Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016333.3 | NP_057417.3 | 11 | 4136 | ||
XM_005255226.1 | XP_005255283.1 | 11 | 4114 | |||
GenBank mRNA | AB002322.3 | - | 11 | 3788 | ||
AB016087.1 | - | 6 | 2958 | |||
AB016090.1 | - | 1 | 966 | |||
AB016092.1 | - | 11 | 3812 | |||
AF201422.1 | - | 11 | 3816 | |||
BX647088.1 | - | 10 | 3682 |